Merck
  • Highly stereoselective double (R)-phenylglycinol-induced cyclocondensation reactions of symmetric aryl bis(oxoacids).
Highly stereoselective double (R)-phenylglycinol-induced cyclocondensation reactions of symmetric aryl bis(oxoacids).
Organic & biomolecular chemistry (2011-02-08)
Mercedes Amat, Carlos Arróniz, Elies Molins, Carmen Escolano, Joan Bosch
ABSTRACT
The double cyclocondensation of symmetric pyridyl bis(oxoacids) 2b and 3b with (R)-phenylglycinol stereoselectively gave access to bis-phenylglycinol-derived oxazolopyrrolidine 9 and oxazolopiperidone 10, respectively. Application of the stereocontrolled cyclocondensation reaction to phenyl bis-γ-oxoacid 4b provided 11, which was converted to the corresponding enantiopure di(pyrrolidinyl)benzene 22. The absolute configuration of the new stereogenic centers generated in the key cyclocondenstion step was unambiguously established by X-ray crystallographic analysis.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%
Pricing and availability is not currently available.