Merck
  • Applications and modifications of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) in peptides and peptidomimetics design and discovery.
Applications and modifications of 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) in peptides and peptidomimetics design and discovery.
Current protein & peptide science (2011-01-18)
Yingjie Zhang, Hao Fang, Wenfang Xu
ABSTRACT
Tic, short for 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, is a kind of unnatural α-amino acids. Due to its distinct geometrical conformation and biological activity, the structure of Tic, regarded as the surrogate of proline and the rigid analogue of phenylalanine or tyrosine, has been introduced into many compounds, which target diverse enzymes or receptors. The most successful example is that substituting the Tic residue for the proline residue of enalapril led to an approved drug quinapril. In this review, we will summarize the applications and modifications of Tic in peptides and peptidomimetics design and discovery, and hope to spark medicinal researchers' inspiration in the field of protein and peptide drug design and optimization.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%