Merck
  • Diastereoselective formal total synthesis of (+/-)-triptolide via a novel cationic cyclization of 2-alkenyl-1,3-dithiolane.
Diastereoselective formal total synthesis of (+/-)-triptolide via a novel cationic cyclization of 2-alkenyl-1,3-dithiolane.
Chemical communications (Cambridge, England) (2010-07-06)
Sylvie Goncalves, Paul Hellier, Marc Nicolas, Alain Wagner, Rachid Baati
ABSTRACT
A concise and diastereoselective formal total synthesis of triptolide, a natural product with a wide range of biological properties, is described. The key reaction is an unprecedented 6-endo-Trig cationic cyclization of a 2-alkenyl-1,3-dithiolane precursor induced by TMSOTf as Lewis acid.
Sigma-Aldrich
1,3-Dithiolane, 97%