Merck
Severe neurotoxicity following ingestion of tetraethyl lead.
Journal of medical toxicology : official journal of the American College of Medical Toxicology (2010-03-23)
Brandon K Wills, Jason Christensen, Joe Mazzoncini, Michael Miller
ABSTRACT
Organic lead compounds are potent neurotoxins which can result in death even from small exposures. Traditionally, these compounds are found in fuel stabilizers, anti-knock agents, and leaded gasoline. Cases of acute organic lead intoxication have not been reported for several decades. We report a case of a 13-year-old Iraqi male who unintentionally ingested a fuel stabilizer containing 80-90% tetraethyl lead, managed at our combat support hospital. The patient developed severe neurologic symptoms including agitation, hallucinations, weakness, and tremor. These symptoms were refractory to escalating doses of benzodiazepines and ultimately required endotracheal intubation and a propofol infusion. Adjunctive therapies included chelation, baclofen, and nutrition provided through a gastrostomy tube. The patient slowly recovered and was discharged in a wheelchair 20 days after ingestion, still requiring tube feeding. Follow-up at 62 days post-ingestion revealed near-resolution of symptoms with residual slurred speech and slight limp. This case highlights the profound neurotoxic manifestations of acute organic lead compounds.