Merck
  • Oral delivery of 1,3-dicyclohexylurea nanosuspension enhances exposure and lowers blood pressure in hypertensive rats.
Oral delivery of 1,3-dicyclohexylurea nanosuspension enhances exposure and lowers blood pressure in hypertensive rats.
Basic & clinical pharmacology & toxicology (2008-03-04)
Sarbani Ghosh, Po-Chang Chiang, Jan L Wahlstrom, Hideji Fujiwara, Jon G Selbo, Steven L Roberds
ABSTRACT
Cytochrome P450-derived epoxyeicosatrienoic acids (EET) are biologically active metabolites of arachidonic acid that have potent effects on renal vascular reactivity and tubular ion transport and have been implicated in the control of blood pressure. EETs are hydrolyzed to their less active diols, dihydroxyeicosatrienoic acids (DHET), by the enzyme soluble epoxide hydrolase (sEH). 1,3-dicyclohexylurea (DCU), a potent sEH inhibitor, lowers systemic blood pressure in spontaneously hypertensive rats when dosed intraperitoneally. However, DCU has poor aqueous solubility, posing a challenge for in vivo oral delivery. To overcome this limitation, we formulated DCU in a nanosuspension using wet milling. Milling reduced particle size, increasing the total surface area by approximately 40-fold. In rats chronically infused with angiotensin II, the DCU nanosuspension administered orally twice daily for 4 days produced plasma exposures an order of magnitude greater than unmilled DCU and lowered blood pressure by nearly 30 mmHg. Consistent with the mechanism of sEH inhibition, DCU increased plasma 14,15-EET and decreased plasma 14,15-DHET levels. These data confirm the antihypertensive effect of sEH inhibition and demonstrate that greatly enhanced exposure of a low-solubility compound is achievable by oral delivery using a nanoparticle drug delivery system.
Sigma-Aldrich
N,N′-Dicyclohexylurea, 98%
Pricing and availability is not currently available.