Merck
[Studies on chemical constituents from root of Clematis hexapetala].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-01-18)
Cai-Xi Dong, She-Po Shi, Ke-Si Wu, Peng-Fei Tu
ABSTRACT
To study the chemical constituents from the roots and rhizomes of Clematis hexapetala. The compounds were separated by means of solvent extraction, repeated chromatography with silica gel and HPLC. The structures were determined by spectral analysis. Nine compounds were isolated as friedelin (1) , anemonin (2) , beta-sitosterol (3) , palmitic acid (4) , vanillic acid (5) , isolariciresinol (6) , 5-hydroxumethyl-5H-furan-2-one (7) , n-nonane (8) , daucosterol (9). All the compounds were isolated from the plant for the first time.