Merck
  • Biological treatment of textile dye acid violet-17 by bacterial consortium in an up-flow immobilized cell bioreactor.
Biological treatment of textile dye acid violet-17 by bacterial consortium in an up-flow immobilized cell bioreactor.
Letters in applied microbiology (2004-04-03)
D K Sharma, H S Saini, M Singh, S S Chimni, B S Chadha
ABSTRACT
To develop a cost effective and efficient biological treatment process for small scale textile processing industries (TPI) releasing untreated effluents containing intense coloured Acid violet-17 (AV-17), a triphenyl methane (TPM) group textile dye. The samples collected from effluent disposal sites of TPI were used for selective enrichment of microbial populations capable of degrading/decolourizing AV-17. A consortium of five bacterial isolates was used to develop an up-flow immobilized cell bioreactor for treatment of feed containing AV-17. The bioreactor, operating at a flow rate of 6 ml x h(-1), resulted in 91% decolourization of 30 mg AV-17/l with 94.3 and 95.7% removal of biochemical oxygen demand and chemical oxygen demand of the feed. Comparison of the input and output of the bioreactor by UV-visible, thin layer chromatography and (1)H-nuclear magnetic resonance spectroscopy indicates conversion of the parent dye into unrelated metabolic intermediates. These results will form a basis for developing 'on-site' treatment system for TPI effluents to achieve decolourization and degradation of residual dyes.
Sigma-Aldrich
Coomassie Violet R200, Dye content 50 %
Pricing and availability is not currently available.