Merck
  • The effects of clofentezine on life-table parameters in two-spotted spider mite Tetranychus urticae.
The effects of clofentezine on life-table parameters in two-spotted spider mite Tetranychus urticae.
Experimental & applied acarology (2004-02-06)
Dejan Marcic
ABSTRACT
Sublethal effects of the growth inhibitor, clofentezine, on life-table parameters of Tetranychus urticae Koch females treated at different developmental stages with a concentration causing >90% mortality were investigated. Females which survived treatment as 'early' (0-24 h old) eggs produced 12% more offspring than the untreated females during the first five days of oviposition. This resulted in a significant rise in the intrinsic rate of increase (rj): 0.324, compared to 0.299 in the untreated females. This effect may be interpreted as hormoligosis. Clofentezine treatment at any other developmental stage of T. urticae significantly decreased both longevity and fertility of female survivors. Females which survived treatment either as 'late' (72-96 h old) eggs or larvae had 2.6 times lower net reproductive rate (R0) than the untreated females, and the rj values were significantly lower: 0.242 and 0.215, respectively (0.285 in the untreated females). Females which survived treatment either as protonymphs or deutonymphs had 3.9 times and 6 times lower R0, respectively. Corresponding rj values were 0.178 and 0.146, respectively (0.247 in the untreated females). The clofentezine treatment at all stages influenced the age distribution of survivors. The sublethal effects of clofentezine and their impact on T. urticae management are discussed.
Product Description
Supelco
Clofentezine, PESTANAL®, analytical standard
Pricing and availability is not currently available.