Merck
  • A stronger patch test elicitation reaction to the allergen hydroxycitronellal plus the irritant sodium lauryl sulfate.
A stronger patch test elicitation reaction to the allergen hydroxycitronellal plus the irritant sodium lauryl sulfate.
Contact dermatitis (2003-12-18)
S Heydorn, K E Andersen, J D Johansen, T Menné
ABSTRACT
Household and cleaning products often contain both allergens and irritants. The aim of this double-blinded, randomized, paired study was to determine whether patch testing with an allergen (hydroxycitronellal) combined with an irritant [sodium lauryl sulfate (SLS)] cause a stronger patch test elicitation reaction than patch testing with the allergen (hydroxycitronellal) alone, in patients previously patch tested positive to hydroxycitronellal. A stronger patch test elicitation reaction was defined as at least 1 day of patch test reading showing more positive patch tests (+, ++ or +++) on the forearm patch tested with 6 concentrations of SLS plus hydroxycitronellal than on the forearm tested with 6 concentrations of hydroxycitronellal alone and no day of patch test readings showing more positive tests on the hydroxycitronellal forearm. 15/20 (75%) had at least 1 day of patch test reading with more positive patch tests on the forearm patch tested with SLS plus hydroxycitronellal and no day of patch test readings with more positive patch tests on the forearm tested with hydroxycitronellal (P = 0.0253). Estimation of the blood flow with laser Doppler supported these findings.
Sigma-Aldrich
Hydroxycitronellal, ≥95%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
7-Hydroxycitronellal, Odorant used in allergy studies
Pricing and availability is not currently available.