Merck
  • Synthesis and analysis of symmetrical and nonsymmetrical disaturated/monounsaturated triacylglycerols.
Synthesis and analysis of symmetrical and nonsymmetrical disaturated/monounsaturated triacylglycerols.
Journal of agricultural and food chemistry (2003-03-20)
Richard O Adlof, Gary R List
ABSTRACT
Symmetrical disaturated triacylglycerols of the structure SUS, where S is stearic acid (18:0) and U is an unsaturated fatty acid, either oleic (O; 9cis-18:1), linoleic (L; 9cis,12cis-18:2), or linolenic (Ln; 9cis,12cis,15cis-18:3), are important components providing functionality to interesterified fat blends and structurally modified oils. Nonsymmetrical triacylglycerols of the structure SSU can significantly change melting point and solid fat content profiles. To characterize the physical properties of pure and symmetrical and nonsymmetrical triacylglycerol mixtures, the same reaction sequence has been used to prepare multigram quantities of triacylglycerols SUS and SSU. Tristearin was converted to a mixture of mono-, di-, and triacylglycerols, and the 1,3- and 1,2-diacylglycerol fraction was isolated by silica column chromatography. The 1,3-diacylglycerols were removed by crystallization from acetone and esterified with the appropriate fatty acid to form the symmetrical triacylglycerols with >99% SUS structure. The more difficult to obtain 1,2-diacylglycerols were prepared by esterification of the enriched 1,2-diacylglycerol fraction (80-86% 1,2-diacylglycerols) remaining after removal of much of the 1,3-isomer by crystallization, but silver resin or silver nitrate impregnated silica gel chromatography was required to isolate the nonsymmetrical triacylglycerols. SSL and SSLn were prepared in purities of >98% by this procedure, but not SSO. Silver ion HPLC was found to be as accurate as, and more rapid than, lipolysis/gas chromatography for the determination of the isomeric purities of the synthesized triacylglycerols.
Sigma-Aldrich
Glyceryl 1,3-distearate, ≥99% (TLC)
Pricing and availability is not currently available.