Merck
  • Solution-phase simultaneous addition of functionalities (SPSAF) and chemical transformation to prepare N,N'-disubstituted piperazine libraries.
Solution-phase simultaneous addition of functionalities (SPSAF) and chemical transformation to prepare N,N'-disubstituted piperazine libraries.
Biotechnology and bioengineering (1999-04-01)
P P Kung, P D Cook
ABSTRACT
Several piperazine libraries were prepared using solution phase simultaneous addition of functionalities methodology as well as the "library from library" concept. The resulting piperazine libraries displayed antimicrobial activity against both gram-positive and gram-negative bacteria.