Merck
  • New aromatic inhibitors of EPSP synthase incorporating hydroxymalonates as novel 3-phosphate replacements.
New aromatic inhibitors of EPSP synthase incorporating hydroxymalonates as novel 3-phosphate replacements.
Bioorganic & medicinal chemistry (1997-02-01)
A Shah, J L Font, M J Miller, J E Ream, M C Walker, J A Sikorski
ABSTRACT
A new, aromatic analogue of the EPSP synthase enzyme reaction intermediate 1 has been identified, which contains a 3-hydroxymalonate moiety in place of the usual 3-phosphate group. This simplified inhibitor was readily prepared in five steps from ethyl 3,4-dihydroxybenzoate. The resulting tetrahedral intermediate mimic 9 is an effective, competitive inhibitor versus S3P with an apparent Ki of 0.57 +/- 0.06 microM. This result demonstrates that 3-hydroxymalonates exhibit potencies comparable to aromatic inhibitors containing the previously identified 3-malonate ether replacements and can thus function as suitable 3-phosphate mimics in this system. These new compounds provide another example in which a simple benzene ring can be used effectively in place of the more complex shikimate ring in the design of EPSP synthase inhibitors. Furthermore, the greater potency of 9 versus the glycolate derivative 10 and the 5-deoxy-analog 11, again confirms the requirement for multiple anionic charges at the dihydroxybenzoate 5-position in order to attain effective inhibition of this enzyme.