Merck
  • Bioconversion of methyl ricinoleate to 4-hydroxy-decanoic acid and to gamma-decalactone by yeasts of the genus Candida.
Bioconversion of methyl ricinoleate to 4-hydroxy-decanoic acid and to gamma-decalactone by yeasts of the genus Candida.
Journal of basic microbiology (1995-01-01)
A Endrizzi, J M Belin
ABSTRACT
The capacity of several strains of yeasts to do the bioconversion of methyl ricinoleate into gamma-decalactone, was studied in a medium containing this methylic ester of fatty acid as sole carbon source. Amongst the strains which are able to do this bioconversion, two types of behaviour are observed: some of the strains produce gamma-decalactone during all the incubation in bioconversion medium while others produce this aroma compound very quickly and then consume it fast too. The tested strains produce at the same time gamma-decalactone and the corresponding acid form (4-hydroxy-decanoic acid), and this, in variable proportions.
Product Description
Supelco
Methyl ricinoleate, analytical standard
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)