Merck
  • Dimetridazole residues in pork tissue. II. Application of liquid chromatographic method to monitor elimination of drug and its major metabolite.
Dimetridazole residues in pork tissue. II. Application of liquid chromatographic method to monitor elimination of drug and its major metabolite.
Journal - Association of Official Analytical Chemists (1988-11-01)
G Carignan, A I MacIntosh, W Skakum, S Sved
ABSTRACT
A study was conducted to monitor the elimination of dimetridazole (DMZ) and its major metabolite 2-hydroxymethyl-1-methyl-5-nitroimidazole (HMMNI) in swine plasma and tissue, using a liquid chromatographic method with electrochemical detector sensitive to 0.5 ppb. The study consisted of 2 experiments. In the preliminary experiment, one young female piglet was fed medicated ration containing 125 ppm dimetridazole (DMZ) for 2 weeks, followed by a withdrawal period using regular ration for 5 days. Another, control, piglet was given regular diet throughout. Plasma concentrations of DMZ and its most important residue, HMMNI, were measured daily at 2 h after the morning feeding and, on days 8 and 15, several times during the day. The 2 h concentrations after 3 days loading ranged from 47 to 77 ppb for DMZ and 424 to 1081 ppb for HMMNI. A daily cycle in the plasma levels was seen for both substances. Upon withdrawal of medication, elimination of drug and metabolite was biexponential with a terminal half-life of 6.7 h. In the second experiment, 5 piglets were medicated as above and slaughtered 2, 6, 12, 25, and 49 h after withdrawal of the medication; the concentration of DMZ and HMMNI was measured in plasma, muscle, kidney, and liver. DMZ in the plasma amounted to 22 and 1.8 ppb at 2 and 6 h, while HMMNI declined from 535 ppb at 2 h to 0.75 ppb at 25 h. Most values for both substances found in muscle were close to those in the plasma; in kidney they amounted to 9-17% of the plasma levels.(ABSTRACT TRUNCATED AT 250 WORDS)
Product Description
Supelco
HMMNI, VETRANAL®, analytical standard
Pricing and availability is not currently available.