Merck
  • 2-Hydroxycarbazole induces Ca2+ release from sarcoplasmic reticulum by activating the ryanodine receptor.
2-Hydroxycarbazole induces Ca2+ release from sarcoplasmic reticulum by activating the ryanodine receptor.
European journal of pharmacology (1998-10-01)
S C Tovey, C L Longland, M Mezna, F Michelangeli
ABSTRACT
2-Hydroxycarbazole was shown to induce Ca2+ release from skeletal muscle and cardiac muscle sarcoplasmic reticulum at concentrations between 100-500 microM. This release was blocked by both 1 mM tetracaine and 30 microM ruthenium red which inhibit the ryanodine receptor or by pre-treatment with 10 mM caffeine which depletes the ryanodine receptor-containing Ca2+ stores. This, in addition to the fact that 2-hydroxycarbazole has little effect on Ca2+ ATPase activity, indicates that it activates Ca2+ release through the ryanodine receptor. The apparent EC50 value for release from both skeletal muscle and cardiac muscle sarcoplasmic reticulum was approximately 200 microM and maximal release occurred at 400-500 microM, making it approximately 20 times more potent than caffeine. The dose-dependency in the extent of Ca2+ release induced by 2-hydroxycarbazole was also apparently highly cooperative for both preparations. That 2-hydroxycarbazole was able to mobilize Ca2+ from non-muscle cell microsomes and in intact TM4 cells (which contain ryanodine receptors), makes this compound a more potent and commercially available alternative to caffeine in studying the role of this intracellular Ca2+ channel in a variety of systems.
Sigma-Aldrich
2-Hydroxycarbazole, 97%
Pricing and availability is not currently available.