Merck
  • Cancer risk from occupational and environmental exposure to polycyclic aromatic hydrocarbons.
Cancer risk from occupational and environmental exposure to polycyclic aromatic hydrocarbons.
Cancer causes & control : CCC (1997-05-01)
P Boffetta, N Jourenkova, P Gustavsson
ABSTRACT
Epidemiologic evidence on the relationship between polycyclic aromatic hydrocarbons (PAH) and cancer is reviewed. High occupational exposure to PAHs occurs in several industries and occupations. Covered here are aluminum production, coal gasification, coke production, iron and steel foundries, tar distillation, shale oil extraction, wood impregnation, roofing, road paving, carbon black production, carbon electrode production, chimney sweeping, and calcium carbide production. In addition, workers exposed to diesel engine exhaust in the transport industry and in related occupations are exposed to PAHs and nitro-PAHs. Heavy exposure to PAHs entails a substantial risk of lung, skin, and bladder cancer, which is not likely to be due to other carcinogenic exposures present in the same industries. The lung seems to be the major target organ of PAH carcinogenicity and increased risk is present in most of the industries and occupations listed above. An increased risk of skin cancer follows high dermal exposure. An increase in bladder cancer risk is found mainly in industries with high exposure to PAHs from coal tars and pitches. Increased risks have been reported for other organs, namely the larynx and the kidney; the available evidence, however, is inconclusive. The results of studies addressing environmental PAH exposure are consistent with these conclusions.
Sigma-Aldrich
Calcium carbide, granulated, technical, ≥75% (gas-volumetric)
Sigma-Aldrich
Calcium carbide, pieces, thickness <10 mm , typically, technical grade, ~80%
Sigma-Aldrich
Calcium carbide, CP