MilliporeSigma
  • Investigation of the mechanism by which ketanserin prolongs the duration of the cardiac action potential.
Investigation of the mechanism by which ketanserin prolongs the duration of the cardiac action potential.
Journal of cardiovascular pharmacology (1995-11-01)
B Le Grand, J M Talmant, J P Rieu, J F Patoiseau, F C Colpaert, G W John
ABSTRACT
Action potential duration (APD) lengthening is believed to underlie the cardiac arrhythmogenicity of ketanserin, a serotonin (5-HT)2A/2C receptor antagonist. We wished to determine (a) whether this activity involves blockade of 5-HT2A/2C receptors and (b) the precise mechanism of ketanserin-induced APD prolongation. APs were recorded in guinea pig isolated papillary muscles by conventional "floating" microelectrodes, and potassium currents in guinea pig isolated myocytes were recorded in the whole-cell configuration. Ketanserin (1-10 microM) increased APD (EC50 value for enhancing APD at 90% repolarization (APD90) 3.1 +/- 2.7 microM, n = 24), without affecting resting potential, maximum upstroke velocity (Vmax) or AP amplitude (APA). Pirenperone (10 microM), a ketanserin congener, similarly increased APD90 from 204 +/- 3 to 241 +/- 7 ms (p < 0.001, n = 6). No increase in APD was observed, however, with ritanserin or ICI 170809, even at high concentrations (10 microM, n = 6, respectively), two 5-HT2A/2C receptor antagonists chemically distinct from ketanserin, thereby excluding the involvement of 5-HT2A/2C receptors in mediating APD lengthening. That APD prolongation was mediated specifically by the benzolyl-piperidine moiety of ketanserin and pirenperone was confirmed by 1-propyl-4(4-fluorobenzoyl)piperidine (PFBP), which evoked APD lengthening effects remarkably similar to those produced by ketanserin and pirenperone (EC50 3.73 +/- 2.6 microM, n = 12). In isolated cardiomyocytes, ketanserin (1-32 microM) selectively and concentration-dependently reduced the IKr component of the delayed outward current (IK) without affecting the inward rectifier current, IK1. Thus, ketanserin (32 microM) significantly reduced IK at a potential value of -20 mV from 813 +/- 65 to 569 +/- 55 pA (p < 0.001, n = 6), whereas at a potential value of -110 mV, IK1 was not significantly affected (730 +/- 103 vs. 603 +/- 143 pA, respectively; n=6). The results demonstrate that APD is prolonged by ketanserin and congeners but not be chemically different 5-HT2A/2C receptor antagonists. The benzoyl-piperidine moiety appears to mediate the APD-prolonging effects of ketanserin and pirenperone specifically. Furthermore, ketanserin-induced APD lengthening does not appear to involve 5-HT2A/2C receptors but is consecutive to direct blockade of myocardial potassium channels.