Merck
  • Effects of adrenocorticotropin on adrenal and plasma 11 beta-hydroxyandrostenedione in the guinea pig and determination of its relative androgen potency.
Effects of adrenocorticotropin on adrenal and plasma 11 beta-hydroxyandrostenedione in the guinea pig and determination of its relative androgen potency.
Steroids (1993-01-01)
B Bélanger, J Fiet, A Bélanger
ABSTRACT
In order to gain a better understanding of adrenal steroid production in guinea pigs, adrenocorticotropin (ACTH) was administered to castrated adult guinea pigs and a series of C-21 and C-19 steroid levels were determined in both adrenals and plasma over a 6-h period. After injection of ACTH, adrenal cortisol and corticosterone concentrations were stimulated by almost 40- and threefold, respectively, whereas the content of C-21 steroid precursors, namely pregnenolone, 17-hydroxypregnenolone, progesterone, and 17-hydroxyprogesterone, was stimulated by two- to 10-fold. Increases of C-19 steroid levels, namely dehydroepiandrosterone (DHEA), androstenedione (4-DIONE), testosterone, and 11 beta-hydroxyandrostenedione (11 beta-DIONE) ranging from 1.5 to eight times were also observed in the adrenals. In castrated guinea pigs, basal plasma DHEA, 4-DIONE, and testosterone levels were undetectable by radioimmunoassay, whereas 11 beta-DIONE concentrations were in a range similar to those in intact animals. In plasma, ACTH had a marked effect on corticosterone, cortisol, and 11 beta-DIONE. The present results indicate that although guinea pig adrenals synthesized several C-19 steroids, only 11 beta-DIONE was secreted by the adrenals into circulation even after administration of ACTH. The biological activity of this hydroxylated C-19 steroid was then assessed by using silastic implants in castrated guinea pigs, and it was shown that plasma 11 beta-DIONE concentrations increased by threefold and had no effect on prostate weight. Moreover, by using ZR-75-1 cells, a mammary cancer cell line extremely sensitive to androgens, we demonstrated that 11 beta-DIONE had an androgenic potency 35-fold less than that of dihydrotestosterone.
Sigma-Aldrich
4-Androsten-11β-ol-3,17-dione