Merck
  • Simple and rapid method for the determination of the diastereomers of difenacoum in blood and liver using high-performance liquid chromatography with fluorescence detection.
Simple and rapid method for the determination of the diastereomers of difenacoum in blood and liver using high-performance liquid chromatography with fluorescence detection.
Journal of chromatography (1993-10-22)
M J Kelly, J Chambers, A D MacNicoll
ABSTRACT
A rapid and sensitive high-performance liquid chromatographic method for the analysis of cis and trans diastereomers of the anticoagulant rodenticide difenacoum has been described. The methodology demonstrates potential for the analysis of diastereomers of related 4-hydroxycoumarin anticoagulants. Separations were achieved by reversed-phase chromatography on a Zorbax ODS column with gradient elution using acetonitrile-water, modified with 0.1% acetic acid, as the mobile phase. Detection of the analytes was effected by fluorescence at excitation and emission wavelengths of 310 and 390 nm, respectively. Sample preparation from both plasma and liver has been simplified to reduce preparation time and manipulation. The minimum detectable concentration of each diastereomer was 5 ng/ml. Recoveries of 100% were obtained from plasma and 93% from liver tissue. This method has been used for the investigation of the pharmacokinetics of difenacoum diastereomers in rats, and for investigation of unexplained hypoprothrombinaemic events encountered clinically.
Product Description
Supelco
Difenacoum, PESTANAL®, analytical standard
Pricing and availability is not currently available.