Merck
Insulinotropic action of glutamic acid dimethyl ester.
The American journal of physiology (1994-10-01)
A Sener, I Conget, J Rasschaert, V Leclercq-Meyer, M L Villanueva-Peñacarrillo, I Valverde, W J Malaisse
ABSTRACT
Glutamic acid dimethyl ester (GME; 3.0-10.0 mM) enhanced insulin release evoked by 6.0-8.3 mM D-glucose, 1.0-10.0 mM L-leucine, or 5.0-10.0 mM 2-amino-bicyclo(2,2,1)heptane-2-carboxylic acid, causing a shift to the left of the sigmoidal relationship between insulin output and D-glucose concentration. In the absence of D-glucose, GME also unmasked the insulinotropic potential of glibenclamide. In islets exposed to L-leucine, the insulinotropic action of GME coincided with an early fall and later increase in 86Rb outflow and augmentation of 45Ca outflow from prelabeled islets. The measurement of O2 uptake, NH4+ output, production of 14CO2 from islets prelabeled with [U-14C]palmitate, generation of 14C-labeled amino acids and 14CO2 from the dimethyl ester of either L-[1-14C]glutamic acid or L-[U-14C]glutamic acid, and D-[2-14C]glucose as well as D-[6-14C]glucose oxidation in the presence or absence of GME indicated that the latter ester was efficiently converted to L-glutamate and its further metabolites. The overall gain in O2 uptake represented the balance between GME oxidation and its sparing action on the catabolism of endogenous fatty acids and exogenous D-glucose. It is proposed that GME might represent a new tool to bypass beta-cell defects in D-glucose transport, phosphorylation, and further metabolism and, hence, to stimulate insulin release in experiments conducted in animal models of non-insulin-dependent diabetes mellitus.
Sigma-Aldrich
L-Glutamic acid dimethyl ester hydrochloride, ≥99.0% (anhydrous basis material, AT)
Pricing and availability is not currently available.