Merck
  • Differing actions of nitropropane analogs of GABA and baclofen in central and peripheral preparations.
Differing actions of nitropropane analogs of GABA and baclofen in central and peripheral preparations.
European journal of pharmacology (1994-10-13)
J Ong, D I Kerr, G Lacey, D R Curtis, R Hughes, R H Prager
ABSTRACT
In the guinea-pig isolated ileum, both baclofen (5-100 microM) and gamma-aminobutyric acid (GABA; 5-100 microM) induced a bicuculline-insensitive depression of cholinergic twitch contractions which was reversibly and competitively antagonised by 3-amino-1-nitropropane (50-250 microM). 3-Amino-1-nitropropane (pA2 = 5.0 +/- 0.1) was twice as potent as 2-hydroxysaclofen (pA2 = 4.5 +/- 0.1), but was equipotent with 3-aminopropyl(P-diethoxymethyl)phosphinic acid (CGP 35348) (pA2 = 4.9 +/- 0.2), and did not show any partial agonist activity at these peripheral GABAA or GABAB receptor sites. In rat neocortical slices, 3-amino-1-nitropropane did not activate GABAB receptor sites or affect baclofen-induced suppression of spontaneous discharges. In the cat spinal cord, however, under in vivo conditions, the corresponding nitro analog of baclofen 3-amino-2-(4-chloro)-nitropropane was an agonist at GABAB receptor sites, although more than 60 times weaker than baclofen in depressing monosynaptic excitatory field potentials, whereas 3-amino-1-nitropropane was an extremely weak agonist at bicuculline-sensitive GABAA sites. The differing actions of 3-amino-1-nitropropane at peripheral and central GABAB receptor sites suggest heterogeneity among these receptors.
Sigma-Aldrich
1-Nitropropane, ≥98.5%
Pricing and availability is not currently available.