Merck
  • Dose-related hepatotoxicity of 1,1,2-trichloro-1,2,2-trifluoroethane in short-term intermittent inhalation exposure in rats.
Dose-related hepatotoxicity of 1,1,2-trichloro-1,2,2-trifluoroethane in short-term intermittent inhalation exposure in rats.
Toxicology (1980-01-01)
H Vainio, J Nickels, T Heinonen
ABSTRACT
Male Wistar rats were exposed to 200, 1000 or 2000 ppm of 1,1,2-trichloro-1,2,2-trifluoroethane vapor 5 days a week 6 h daily for 1 or 2 weeks. Proliferation and vacuolisation of the smooth endoplasmic reticulum (SER) of the liver was seen electron microscopically after 1 and 2 weeks in the rats exposed to 1000 and 2000 ppm. Among the hepatic drug metabolizing enzymes, NADPH cytochrome c reductase activity showed a dose-related decrease whereas the tightly membrane-bound UDPglucuronosyltransferase exhibited a dose-dependent enhancement in its measurable activity. The overall drug oxidation reaction, 7-ethoxycoumarin O-deethylase was not affected by the 1,1,2-trichloro-1,2,2,-trifluoroethane inhalation at all, either in the liver or in the kidneys. 1,1,2-Trichloro-1,2,2-trifluoroethane binds to cytochrome P-450 with the production of a type I difference spectrum, suggesting that it may act as a substrate for this enzyme. The binding affinity is increased by phenobarbital-treatment of the rats.