Merck
  • Herpes virus infection and uterine cervix carcinoma. General considerations, personal experimental data and suggestions for a specific antiherpes adjuvant therapy.
Herpes virus infection and uterine cervix carcinoma. General considerations, personal experimental data and suggestions for a specific antiherpes adjuvant therapy.
Virologie (1982-04-01)
E Nastac, M Hozoc, M Stoian
ABSTRACT
The authors review some data of the literature and results of personal investigations that point out a relationship between human herpes simplex virus infection and the etiopathogeny of uterine cervix carcinoma. On the ground of these data the authors suggest a "specific antiherpes" adjuvant therapy in this form of human cancer.
Sigma-Aldrich
Moroxydine hydrochloride, 99%