Merck
  • Relationship between membrane fluidity and capping of receptors for concanavalin A.
Relationship between membrane fluidity and capping of receptors for concanavalin A.
Canadian journal of biochemistry (1980-12-01)
W W Mak, J T Wong
ABSTRACT
The activities of a range of phenylalaninol-related compounds on capping of concanavalin A and induction of rounding of Chinese hamster ovary tsHl cells, as well as on the fluidity of phosphatidylcholine-cholesterol (1:1) liposomes, have been examined. These compounds include phenylalaninol, histidinol, leucinol, benzyl alcohol, benzylamine, 2-phenylethanol, 2-phenylamine, 3-phenyl-1-propanol, 3-phenyl-1-propylamine, and 3-phenylpropionic acid. The results indicate a strong correlation between the capacities of these compounds to enhance fluidity and their capacities to inhibit capping of concanavalin A. The specificity of this correlation is suggested by the finding that both types of capacities are poorly correlated with the capacities of the various compounds to induce cell rounding.
Sigma-Aldrich
(S)-(−)-2-Amino-3-phenyl-1-propanol, 98%, optical purity ee: 99% (HPLC)
Pricing and availability is not currently available.