Merck
A rapid, sensitive myeloperoxidase stain using 4-chloro-1-naphthol.
American journal of clinical pathology (1980-06-01)
J M Elias
ABSTRACT
The substitution of 4-chloro-1-naphthol for benzidine-containing compounds used in the myeloperoxidase stain for distinguishing granulocytes from lymphoid blood elements is described. 4-Chloro-1-naphthol is an acceptable substitute for benzidine compounds, which are considered to be carcinogenic. The feasibility of using this method on peripheral blood and bone-marrow films is discussed.
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
4-Chloro-1-naphthol, tablet
Pricing and availability is not currently available.