Merck
  • Effect of substrate and pH on the oxidative half-reaction of phenol hydroxylase.
Effect of substrate and pH on the oxidative half-reaction of phenol hydroxylase.
The Journal of biological chemistry (1985-05-25)
K Detmer, V Massey
ABSTRACT
The oxidative half-reaction of phenol hydroxylase has been studied by stopped-flow spectrophotometry. Three flavin-oxygen intermediates can be detected when the substrate is thiophenol, or m-NH2, m-OH, m-CH3, m-Cl, or p-OH phenol. Intermediate I, the flavin C(4a)-hydroperoxide, has an absorbance maximum at 380-390 nm and an extinction coefficient approximately 10,000 M-1 cm-1. Intermediate III, the flavin C(4a)-hydroxide, has an absorbance maximum at 365-375 nm and an extinction coefficient approximately 10,000 M-1 cm-1. Intermediate II has absorbance maxima of 350-390 nm and extinction coefficients of 10,000-16,000 M-1 cm-1 depending on the substrate. A Hammett plot of the logarithm of the rates of the oxygen transfer step, the conversion of intermediate I to intermediate II, gives a straight line with a slope -0.5. Fluoride ion is a product of the enzymatic reaction when 2,3,5,6-tetrafluorophenol is the substrate. These results are consistent with an electrophilic substitution mechanism for oxygen transfer. The conversions of I to II and II to III are acid-catalyzed. A kinetic isotope effect of 8 was measured for the conversion of II to III using deuterated resorcinol as substrate. The conversion of III to oxidized enzyme is base-catalyzed, suggesting that the reaction depends on the removal of the flavin N(5) proton. Product release occurs at the same time as the formation of intermediate III, or rapidly thereafter. The results are interpreted according to the ring-opened model of Entsch et al. (Entsch, B., Ballou, D. P., and Massey, V. (1976) J. Biol. Chem. 251, 2550-2563).
Sigma-Aldrich
2,3,5,6-Tetrafluorophenol, 97%
Pricing and availability is not currently available.