Merck
Isolation and properties of beta-glucosidase from Ruminococcus albus.
Journal of bacteriology (1985-01-01)
K Ohmiya, M Shirai, Y Kurachi, S Shimizu
ABSTRACT
An enzyme active against p-nitrophenyl-beta-D-glucoside was purified from logarithmic-phase cells of Ruminococcus albus cultivated in a medium containing ball-milled cellulose. The purification yielded homogeneous enzyme after an approximately 520-fold increase in specific activity and a 9% yield. The enzyme was identified as a beta-glucosidase because it can hydrolyze cellobiose and cellooligosaccharides to glucose from the nonreducing ends.
Sigma-Aldrich
4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)