Merck
  • [Chemico-toxicologic findings in fatal suicidal poisoning by demeton-S-methyl].
[Chemico-toxicologic findings in fatal suicidal poisoning by demeton-S-methyl].
Zeitschrift fur Rechtsmedizin. Journal of legal medicine (1988-01-01)
D Schludecker, R Aderjan
ABSTRACT
A case of lethal suicidal intoxication with demeton-S-methyl is reported. Capillary chromatography on wide-bore columns (CP-sil 5, 0.53 mm ID, 1 micron film thickness) was used for the quantitative determinations of this substance in the body fluids and tissues.
Product Description
Supelco
Demeton-S-methyl solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.