Merck
  • Multinuclear magnetic resonance studies on serine protease transition state analogues.
Multinuclear magnetic resonance studies on serine protease transition state analogues.
Journal of cellular biochemistry (1989-06-01)
F Adebodun, F Jordan
ABSTRACT
31P Nuclear Magnetic Resonance (NMR) studies were performed on mono- and diisopropylphosphoryl derivatives of alpha-chymotrypsin, trypsin, and subtilisin. Questions addressed included the pKa of the active center Asp...His...Ser triad in both species. While the pKa in the diisopropylphosphoryl derivatives is near 7.4 (found in this and other laboratories earlier) and reflects a nearly normal imidazolium titration curve, the apparent pKa in the monoisopropylphosphoryl enzymes (obtained by "aging" of the diisopropylphosphoryl derivatives and monitored by 31P NMR) is between 9.7 and 11.4 depending on the protease. This latter "titration" of the 31P NMR signal is reversible and presumably reflects the interaction of the imidazolium positive charge with the monoanionic phosphodiester. Of the two tetrahedral intermediates, the properties of the monoisopropylphosphoryl enzyme are probably more representative of the tetrahedral oxyanionic intermediate invoked during peptide hydrolysis. The same NMR technique was used to determine the action of PAM (pyridine-2-aldoxime methiodide, a known "antidote" for acetylcholinesterase inactivated by diisopropylfluorophosphate), on the inactivated enzymes. It was clear that the "antidote" could reverse the diisopropylphosphorylation but was ineffective on the monoisopropylphosphoryl ("aged") enzyme. 11B NMR studies were performed on phenylboronic (PBA) acid and 3,5-bis-trifluoromethylphenylboronic acid in the absence and presence of chymotrypsin and subtilisin. At 22 degrees C the former, but not the latter, compound was in fast exchange between the free and enzyme bound states. The relaxation parameters could be calculated for the bound PBA in chymotrypsin and the fluorinated analogue in subtilisin and clearly indicated that the boron nucleus was tetrahedral in the active centers, a good analogue for the tetrahedral oxyanionic intermediate.
Sigma-Aldrich
3,5-Bis(trifluoromethyl)phenylboronic acid, ≥95%
Pricing and availability is not currently available.