Merck
  • Association between a genetic variant related to glutamic acid metabolism and coronary heart disease in individuals with type 2 diabetes.
Association between a genetic variant related to glutamic acid metabolism and coronary heart disease in individuals with type 2 diabetes.
JAMA (2013-08-29)
Lu Qi, Qibin Qi, Sabrina Prudente, Christine Mendonca, Francesco Andreozzi, Natalia di Pietro, Mariella Sturma, Valeria Novelli, Gaia Chiara Mannino, Gloria Formoso, Ernest V Gervino, Thomas H Hauser, Jochen D Muehlschlegel, Monika A Niewczas, Andrzej S Krolewski, Gianni Biolo, Assunta Pandolfi, Eric Rimm, Giorgio Sesti, Vincenzo Trischitta, Frank Hu, Alessandro Doria
ABSTRACT
Diabetes is associated with an elevated risk of coronary heart disease (CHD). Previous studies have suggested that the genetic factors predisposing to excess cardiovascular risk may be different in diabetic and nondiabetic individuals. To identify genetic determinants of CHD that are specific to patients with diabetes. We studied 5 independent sets of CHD cases and CHD-negative controls from the Nurses' Health Study (enrolled in 1976 and followed up through 2008), Health Professionals Follow-up Study (enrolled in 1986 and followed up through 2008), Joslin Heart Study (enrolled in 2001-2008), Gargano Heart Study (enrolled in 2001-2008), and Catanzaro Study (enrolled in 2004-2010). Included were a total of 1517 CHD cases and 2671 CHD-negative controls, all with type 2 diabetes. Results in diabetic patients were compared with those in 737 nondiabetic CHD cases and 1637 nondiabetic CHD-negative controls from the Nurses' Health Study and Health Professionals Follow-up Study cohorts. Exposures included 2,543,016 common genetic variants occurring throughout the genome. Coronary heart disease--defined as fatal or nonfatal myocardial infarction, coronary artery bypass grafting, percutaneous transluminal coronary angioplasty, or angiographic evidence of significant stenosis of the coronary arteries. A variant on chromosome 1q25 (rs10911021) was consistently associated with CHD risk among diabetic participants, with risk allele frequencies of 0.733 in cases vs 0.679 in controls (odds ratio, 1.36 [95% CI, 1.22-1.51]; P = 2 × 10(-8)). No association between this variant and CHD was detected among nondiabetic participants, with risk allele frequencies of 0.697 in cases vs 0.696 in controls (odds ratio, 0.99 [95% CI, 0.87-1.13]; P = .89), consistent with a significant gene × diabetes interaction on CHD risk (P = 2 × 10(-4)). Compared with protective allele homozygotes, rs10911021 risk allele homozygotes were characterized by a 32% decrease in the expression of the neighboring glutamate-ammonia ligase (GLUL) gene in human endothelial cells (P = .0048). A decreased ratio between plasma levels of γ-glutamyl cycle intermediates pyroglutamic and glutamic acid was also shown in risk allele homozygotes (P = .029). A single-nucleotide polymorphism (rs10911021) was identified that was significantly associated with CHD among persons with diabetes but not in those without diabetes and was functionally related to glutamic acid metabolism, suggesting a mechanistic link.
Product Description
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid hydrochloride
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine Synthetase from Escherichia coli, lyophilized powder, 400-2,000 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, FCC
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SKU
Pack Size
Availability
Price
Quantity
SAFC
L-Glutamine