Merck
  • [Preparation of dealcholized onetaxotere®, docetaxel injection formulation and its pharmacokinetics in rats].
[Preparation of dealcholized onetaxotere®, docetaxel injection formulation and its pharmacokinetics in rats].
Gan to kagaku ryoho. Cancer & chemotherapy (2013-07-19)
Katsuhiro Onishi, Ikuto Tsukiyama, Miki Katoh, Jun Ueyama, Fumie Abe, Hiroko Saito, Katsuhiko Matsuura, Takaaki Hasegawa, Masayuki Nadai
ABSTRACT
Onetaxotere®(OTAX)injection, which is a docetaxel(DOC)injection formulation, cannot be administered to those patients with alcohol intolerance or hypersensitivity, because it contains ethanol as a dissolving agent. To broaden treatment options for those patients, we tried to eliminate ethanol from OTAX injection. Under sterile conditions, dealcoholization was carried out using nitrogen gas in a hot water bath at 50°C. By this method, the ethanol included in OTAX injection was almost completely removed and DOC in the formulation was stable for 28 days. When the dealcoholized or untreated OTAX injection was intravenously injected in rats, no significant differences in the pharmacokinetic parameters of DOC were observed between those with dealcoholized and untreated OTAX injections. It is expected that dealcoholized OTAX may be useful in patients with alcohol-related difficulties.
Sigma-Aldrich
Reagent Alcohol, reagent grade
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Alcohol dehydrated, ≥85.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ethanol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis