Merck
Cadmium exposure induces hematuria in Korean adults.
Environmental research (2013-05-07)
Seung Seok Han, Myounghee Kim, Su Mi Lee, Jung Pyo Lee, Sejoong Kim, Kwon Wook Joo, Chun Soo Lim, Yon Su Kim, Dong Ki Kim
ABSTRACT
Toxic heavy metals have adverse effects on human health. However, the risk of hematuria caused by heavy metal exposure has not been evaluated. Data from 4701 Korean adults were obtained in the Korean National Health and Nutritional Examination Survey (2008-2010). Blood levels of the toxic heavy metals cadmium, lead, and mercury were measured. Hematuria was defined as a result of ≥+1 on a urine dipstick test. The odds ratios (ORs) for hematuria were measured according to the blood heavy metal levels after adjusting for multiple variables. Individuals with blood cadmium levels in the 3rd and 4th quartiles had a greater OR for hematuria than those in the 1st quartile group: 3rd quartile, 1.35 (1.019-1.777; P=0.037); 4th quartile, 1.52 (1.140-2.017; P=0.004). When blood cadmium was considered as a log-transformed continuous variable, the correlation between blood cadmium and hematuria was significant: OR, 1.97 (1.224-3.160; Ptrend=0.005). In contrast, no significant correlations between hematuria and blood lead or mercury were found in the multivariate analyses. The present study shows that high cadmium exposure is associated with a risk of hematuria.
Sigma-Aldrich
Mercury, SAJ first grade, ≥99.5%
Sigma-Aldrich
Mercury, JIS special grade, ≥99.5%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis