Merck
  • Enantioselective synthesis of pactamycin, a complex antitumor antibiotic.
Enantioselective synthesis of pactamycin, a complex antitumor antibiotic.
Science (New York, N.Y.) (2013-04-13)
Justin T Malinowski, Robert J Sharpe, Jeffrey S Johnson
ABSTRACT
Medicinal application of many complex natural products is precluded by the impracticality of their chemical synthesis. Pactamycin, the most structurally intricate aminocyclopentitol antibiotic, displays potent antiproliferative properties across multiple phylogenetic domains, but it is highly cytotoxic. A limited number of analogs produced by genetic engineering technologies show reduced cytotoxicity against mammalian cells, renewing promise for therapeutic applications. For decades, an efficient synthesis of pactamycin amenable to analog derivatizations has eluded researchers. Here, we present a short asymmetric total synthesis of pactamycin. An enantioselective Mannich reaction and symmetry-breaking reduction sequence was designed to enable assembly of the entire carbon core skeleton in under five steps and control critical three-dimensional (stereochemical) functional group relationships. This modular route totals 15 steps and is immediately amenable for structural analog synthesis.
Sigma-Aldrich
3′-Aminoacetophenone, 97%
Pricing and availability is not currently available.