Merck
  • An ire1-phk1 chimera reveals a dispensable role of autokinase activity in endoplasmic reticulum stress response.
An ire1-phk1 chimera reveals a dispensable role of autokinase activity in endoplasmic reticulum stress response.
Journal of molecular biology (2013-04-02)
M Amin-ul Mannan, William R Shadrick, Gabriel Biener, Byung-Sik Shin, Ashish Anshu, Valerica Raicu, David N Frick, Madhusudan Dey
ABSTRACT
The endoplasmic reticulum transmembrane receptor Ire1 senses over-accumulation of unfolded proteins in the endoplasmic reticulum and initiates the unfolded protein response (UPR). The cytoplasmic portion of Ire1 has a protein kinase domain (KD) and a kinase extension nuclease (KEN) domain that cleaves an mRNA for encoding the Hac1 transcription factor needed to express UPR genes. During this UPR signaling, Ire1 proteins self-assemble into an oligomer of dimers, which essentially requires autophosphorylation of a constituent activation loop in the KD. However, it is not clear how dimerization, autophosphorylation, and KEN domain function are precisely coordinated. In this study, we uncoupled the KD and KEN domain functions, by removing the activation loop along with an extended region that we called the auto-inhibitory region (AIR), or by swapping the activation loop with a homologous loop from phosphorylase kinase 1 (Ire1(PHK)). Both Ire1(ΔAIR) and Ire1(PHK) activated the UPR even when either protein contained a mutation (D797A) that abolished the ability of Ire1 KD to transfer phosphates to the AIR. Neither protein functioned when containing mutations in key ATP binding residues (E746A and N749A) or in residues that disrupted Ire1 dimer interface (W426A or R697D). We interpret these results as evidence supporting the notion that the primary function of the kinase domain is to autophosphorylate the AIR in order to relieve auto-inhibition and that ADP acts as a switch to activate the KEN domain-catalyzed HAC1 mRNA cleavage.
Sigma-Aldrich
Phosphorylase Kinase from rabbit muscle, lyophilized powder, ≥60 units/mg protein
Pricing and availability is not currently available.