Merck
  • Amitriptyline for symptomatic treatment of idiopathic chronic laryngeal irritability.
Amitriptyline for symptomatic treatment of idiopathic chronic laryngeal irritability.
The Annals of otology, rhinology, and laryngology (2013-03-12)
Daniel J Stein, J Pieter Noordzij
ABSTRACT
We evaluated the safety and efficacy of amitriptyline hydrochloride in treating idiopathic chronic laryngeal irritability. A retrospective chart review identified patients treated with amitriptyline for laryngeal irritability in an academic otolaryngology practice. Subjects who had documented medication compliance and a follow-up evaluation performed within 3 months after initiation of treatment were included. Symptoms, demographic information, and response to therapy were abstracted from the records. Sixty-six subjects were identified, comprising 48 women and 18 men. The ethnicities were black, 42%; Hispanic, 27%; white, 20%; Asian, 6%; and other or unspecified, 5%. The response to treatment was judged complete in 32% of subjects, partial in 24%, and without improvement in 36%; the remaining 8% were unable to tolerate treatment. Overall, 56.1% of subjects improved (95% confidence limit, 43.3% to 68.3%). No significant differences in response rates were seen based on gender (p = 0.484), age (p = 0.590), or race (p = 0.846). Sedation was reported by 23% of individuals. We found that more than 50% of subjects who received amitriptyline for chronic laryngeal irritability experienced improvement, and most subjects tolerated this treatment. No differences in efficacy were seen among racial, age, and gender subgroups. A prospective randomized trial of this therapy appears warranted.
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Pricing and availability is not currently available.
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.