Merck
  • Highly efficient production of L-lactic acid from xylose by newly isolated Bacillus coagulans C106.
Highly efficient production of L-lactic acid from xylose by newly isolated Bacillus coagulans C106.
Bioresource technology (2013-02-13)
Lidan Ye, Xingding Zhou, Mohammad Sufian Bin Hudari, Zhi Li, Jin Chuan Wu
ABSTRACT
Cost-effective production of optically pure lactic acid from lignocellulose sugars is commercially attractive but challenging. Bacillus coagulans C106 was isolated from environment and used to produce l-lactic acid from xylose at 50°C and pH 6.0 in mineral salts medium containing 1-2% (w/v) of yeast extract without sterilizing the medium before fermentation. In batch fermentation with 85g/L of xylose, lactic acid titer and productivity reached 83.6g/L and 7.5g/Lh, respectively. When fed-batch (120+80+60g/L) fermentation was applied, they reached 215.7g/L and 4.0g/Lh, respectively. In both cases, the lactic acid yield and optical purity reached 95% and 99.6%, respectively. The lactic acid titer and productivity on xylose are the highest among those ever reported. Ca(OH)2 was found to be a better neutralizing agent than NaOH in terms of its giving higher lactic acid titer (1.2-fold) and productivity (1.8-fold) under the same conditions.
Sigma-Aldrich
Calcium hydroxide, SAJ special grade, ≥96.0%
Sigma-Aldrich
Calcium hydroxide, CP
Sigma-Aldrich
Calcium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%