Merck
  • Antifungal coatings based on Ca(OH)2 mixed with ZnO/TiO2 nanomaterials for protection of limestone monuments.
Antifungal coatings based on Ca(OH)2 mixed with ZnO/TiO2 nanomaterials for protection of limestone monuments.
ACS applied materials & interfaces (2013-01-26)
Nikte Gómez-Ortíz, Susana De la Rosa-García, William González-Gómez, Montserrat Soria-Castro, Patricia Quintana, Gerko Oskam, Benjamin Ortega-Morales
ABSTRACT
The presence and deteriorating action of microbial biofilms on historic stone buildings have received considerable attention in the past few years. Among microorganisms, fungi are one of the most damaging groups. In the present work, antimicrobial surfaces were prepared using suspensions of Ca(OH)2 particles, mixed with ZnO or TiO2 nanoparticles. The antimicrobial surfaces were evaluated for their antifungal activity both in the dark and under simulated natural photoperiod cycles, using Penicillium oxalicum and Aspergillus niger as model organisms, and two limestone lithotypes commonly used in construction and as materials for the restoration of historic buildings. Both Ca(OH)2-ZnO and Ca(OH)2-TiO2 materials displayed antifungal activity: ZnO-based systems had the best antifungal properties, being effective both in the dark and under illumination. In contrast, TiO2-based coatings showed antifungal activity only under photoperiod conditions. Controls with coatings consisting of only Ca(OH)2 were readily colonized by both fungi. The antifungal activity was monitored by direct observation with microscope, X-ray diffraction (XRD), and scanning electron microscopy (SEM), and was found to be different for the two lithotypes, suggesting that the mineral grain distribution and porosity played a role in the activity. XRD was used to investigate the formation of biominerals as indicator of the fungal attack of the limestone materials, while SEM illustrated the influence of porosity of both the limestone material and the coatings on the fungal penetration into the limestone. The coated nanosystems based on Ca(OH)2-50%ZnO and pure zincite nanoparticulate films have promising performance on low porosity limestone, showing good antifungal properties against P. oxalicum and A. niger under simulated photoperiod conditions.
Sigma-Aldrich
Calcium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Calcium hydroxide, CP
Sigma-Aldrich
Calcium hydroxide, SAJ special grade, ≥96.0%
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis