Merck
  • A new dibenz[b,e]oxepine derivative, 1-hydroxy-10-methoxy-dibenz[b,e]oxepin-6,11-dione, from a marine-derived fungus, Beauveria bassiana TPU942.
A new dibenz[b,e]oxepine derivative, 1-hydroxy-10-methoxy-dibenz[b,e]oxepin-6,11-dione, from a marine-derived fungus, Beauveria bassiana TPU942.
Marine drugs (2013-01-25)
Hiroyuki Yamazaki, Henki Rotinsulu, Tsuyoshi Kaneko, Kazuki Murakami, Hiromu Fujiwara, Kazuyo Ukai, Michio Namikoshi
ABSTRACT
1-Hydroxy-10-methoxy-dibenz[b,e]oxepin-6,11-dione (1) was obtained from the culture broth of a marine-derived fungus, Beauveria bassiana TPU942, isolated from a marine sponge collected at Iriomote Island in Okinawa, together with two known compounds, chrysazin (2) and globosuxanthone A (3). The structure of 1 was elucidated on the basis of its spectroscopic data (HREIMS, 1D and 2D NMR experiments including ¹H--¹H COSY, HMQC and HMBC spectra). Dibenz[b,e]oxepines are rare in nature, and only six natural products have been reported. Therefore, compound 1 is the seventh natural product in this class. Compounds 2 and 3 showed an antifungal activity against Candida albicans, and 3 inhibited the cell growth against two human cancer cell lines, HCT-15 (colon) and Jurkat (T-cell lymphoma). Compound 1 did not show an apparent activity in the same bioassays.