Merck
  • Clinical outcomes of triamcinolone-assisted anterior vitrectomy after phacoemulsification complicated by posterior capsule rupture.
Clinical outcomes of triamcinolone-assisted anterior vitrectomy after phacoemulsification complicated by posterior capsule rupture.
Journal of cataract and refractive surgery (2013-01-23)
Shivani Kasbekar, Som Prasad, Balakrishna Vineeth Kumar
ABSTRACT
To compare the clinical outcomes in patients who had triamcinolone acetate-assisted anterior vitrectomy and patients who had anterior vitrectomy without triamcinolone acetate after phacoemulsification complicated by posterior capsule rupture and vitreous loss. Arrowe Park Hospital, Wirral, United Kingdom. Retrospective consecutive case note review. Consecutive case notes of patients who had anterior vitrectomy assisted by triamcinolone acetonide (triamcinolone group) or without triamcinolone acetate (no-triamcinolone group) after posterior capsule rupture between January 2007 and January 2011 were identified and examined. Data recorded at the clinic visit preoperatively and 1 day and 3 months postoperatively were collated. Information recorded on the pro forma included visual acuity, ocular comorbidities, intraocular pressure (IOP), vitreous strands in the anterior chamber, and other adverse events. No statistically significant difference was found in the visual acuity or IOP between 17 patients in the triamcinolone group and 34 patients in the no-triamcinolone group at any time point. Vitreous strands in the anterior chamber were noted in 1 patient in the triamcinolone group and 7 patients in the no-triamcinolone group. Cystoid macular edema (CME) was present in 3 patients in the no-triamcinolone group, including 1 patient with vitreomacular traction. There was no significant increase in IOP after triamcinolone acetate-assisted anterior vitrectomy. Higher rates of CME and residual anterior chamber vitreous strands in the no-triamcinolone acetate group support the clinical use of triamcinolone acetate.
Product Description
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Triamcinolone acetonide, analytical standard
Pricing and availability is not currently available.