Merck
  • Mitochondrial calcium uniporter blocker effectively prevents brain mitochondrial dysfunction caused by iron overload.
Mitochondrial calcium uniporter blocker effectively prevents brain mitochondrial dysfunction caused by iron overload.
Life sciences (2013-01-22)
Jirapas Sripetchwandee, Jantira Sanit, Nipon Chattipakorn, Siriporn C Chattipakorn
ABSTRACT
Although iron overload induces oxidative stress and brain mitochondrial dysfunction, and is associated with neurodegenerative diseases, brain mitochondrial iron uptake has not been investigated. We determined the role of mitochondrial calcium uniporter (MCU) in brain mitochondria as a major route for iron entry. We hypothesized that iron overload causes brain mitochondrial dysfunction, and that the MCU blocker prevents iron entry into mitochondria, thus attenuating mitochondrial dysfunction. Isolated brain mitochondria from male Wistar rats were used. Iron (Fe(2+) and Fe(3+)) at 0-286 μM were applied onto mitochondria at various incubation times (5-30 min), and the mitochondrial function was determined. Effects of MCU blocker (Ru-360) and iron chelator were studied. Both Fe(2+) and Fe(3+) entered brain mitochondria and caused mitochondrial swelling in a dose- and time-dependent manner, and caused mitochondrial depolarization and increased ROS production. However, Fe(2+) caused more severe mitochondrial dysfunction than Fe(3+). Although all drugs attenuated mitochondrial dysfunction caused by iron overload, only an MCU blocker could completely prevent ROS production and mitochondrial depolarization. Our findings indicated that iron overload caused brain mitochondrial dysfunction, and that an MCU blocker effectively prevented this impairment, suggesting that MCU could be the major portal for brain mitochondrial iron uptake.
Sigma-Aldrich
Ammonium iron(III) citrate, CP, green
Sigma-Aldrich
Ammonium iron(III) citrate, reagent grade, powder
Millipore
Fraser Supplement, suitable for microbiology