Merck
  • Photoaffinity labeling of the sigma-1 receptor with N-[3-(4-nitrophenyl)propyl]-N-dodecylamine: evidence of receptor dimers.
Photoaffinity labeling of the sigma-1 receptor with N-[3-(4-nitrophenyl)propyl]-N-dodecylamine: evidence of receptor dimers.
Biochemistry (2013-01-18)
Uyen B Chu, Subramaniam Ramachandran, Abdol R Hajipour, Arnold E Ruoho
ABSTRACT
The sigma-1 receptor is a ligand-regulated endoplasmic reticulum (ER) resident chaperone involved in the maintenance of cellular homeostasis. Coupling of the sigma-1 receptor with various ER and/or plasma membrane ion channels is associated with its ability to regulate the locomotor activity and cellular proliferation produced in response to sigma-1 receptor ligands. A number of endogenous small molecules bind to the sigma-1 receptor and have been shown to regulate its activity; these include progesterone, N,N-dimethyltryptamine, d-erythro-sphingosine, and/or other endogenous lipids. We previously reported the synthesis of long chain N-alkylamine derivatives and the characterization of the structure-activity relationship between the chain length of N-alkylamine and affinities at the sigma-1 receptor. Here, we present data demonstrating the photoincorporation of one of these N-alkylamine derivatives, N-[3-(4-nitrophenyl)propyl]-N-dodecylamine (4-NPPC12), to the sigma-1 receptor. Matrix-assisted laser desorption ionization time-of-flight and tandem mass spectrometry showed that 4-NPPC12 photoinserted at histidine 154 of the derivatized population of the sigma-1 receptor. Interestingly, light-dependent photoinsertion of 4-NPPC12 resulted in an enhanced electrophoretic mobility of only 50% of the derivatized receptor molecules as assessed by sodium dodecyl sulfate-polyacrylamide gel electrophoresis. The proposed binding and reactivity of 4-NPPC12 evoke a ligand binding model for the sigma-1 receptor that likely involves a receptor dimer and/or oligomer.
Sigma-Aldrich
Dodecylamine, puriss., ≥99.5% (GC)
Sigma-Aldrich
Dodecylamine, ≥99%
Sigma-Aldrich
Dodecylamine, 98%