Merck
  • Reduction of 4-ethylphenol production in red wines using HCDC+ yeasts and cinnamyl esterases.
Reduction of 4-ethylphenol production in red wines using HCDC+ yeasts and cinnamyl esterases.
Enzyme and microbial technology (2013-01-01)
A Morata, R Vejarano, G Ridolfi, S Benito, F Palomero, C Uthurry, W Tesfaye, C González, J A Suárez-Lepe
ABSTRACT
Hydroxycinnamate decarboxylase (HCDC) activity has been evaluated in several commercial yeast strains. The combined effect of using cinnamyl esterases (CE) and HCDC+ Saccharomyces cerevisiae strains has been studied in the formation of vinylphenolic pyranoanthocyanins (VPAs) during fermentation, analysing the kind and concentration of pigments formed according to the yeast strain used. Wines fermented with yeasts HCDC+ were contaminated with Dekkera bruxellensis and afterwards analysed to evaluate the formation of ethylphenols (EPs). The musts treated with CE and later fermented with HCDC+ yeast strains showed lower contents of 4-ethylphenol than those fermented with HCDC- strains. This reduction in the EP content is due to the transformation of hydroxycinnamic acids in stable VPAs pigments. The associated use of CEs and HCDC+ Saccharomyces strains is a natural strategy to reduce the formation of EPs in wines contaminated by Dekkera/Brettanomyces.
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Ethylphenol, 99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
trans-p-Coumaric acid, analytical standard