Merck
  • Iron sulfide catalyzed redox/condensation cascade reaction between 2-amino/hydroxy nitrobenzenes and activated methyl groups: a straightforward atom economical approach to 2-hetaryl-benzimidazoles and -benzoxazoles.
Iron sulfide catalyzed redox/condensation cascade reaction between 2-amino/hydroxy nitrobenzenes and activated methyl groups: a straightforward atom economical approach to 2-hetaryl-benzimidazoles and -benzoxazoles.
Journal of the American Chemical Society (2012-12-20)
Thanh Binh Nguyen, Ludmila Ermolenko, Ali Al-Mourabit
ABSTRACT
Iron sulfide generated in situ from elemental sulfur and iron was found to be highly efficient in catalyzing a redox/condensation cascade reaction between 2-amino/hydroxy nitrobenzenes and activated methyl groups. This method represents a straightforward and highly atom economical approach to 2-hetaryl-benzimidazoles and -benzoxazoles.
Sigma-Aldrich
Iron(II) sulfide, SAJ first grade, ≥50.0%
Sigma-Aldrich
Iron(II) sulfide, technical grade
Sigma-Aldrich
Iron(II) sulfide, −100 mesh, 99.9% trace metals basis