Merck
  • [Modified montmorillonite as multifunction gene and drug delivery system].
[Modified montmorillonite as multifunction gene and drug delivery system].
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences (2012-12-15)
Xue Jin, Qi-wen Wang, Tian-nan Hu, Sang Xu, Qi-da Hu, Jun Zhou, Gu-ping Tang, Xiu-rong Hu
ABSTRACT
To develop polyethylenimine-Doxorubicin-montmorillonite (PEI-Dox-MTT) as a novel multifunction delivery system. Dox was intercalated into montmorillonite, PEI covered to the surface of Dox/MMT to make the nano-particle. XRD, FT-IR and TGA were used to confirm chemical property of the nano-particle. SEM was used to observe the morphology. The capability of drug release was investigated by PBS buffer solution (pH 7.4). The DNA binding ability of nano-particle was detected by gel electrophoresis retardation assay. The cell viability in COS-7 and SKOV3 cell lines was tested using MTT assay. The gastric mucosa protection was evaluated in vitro. XRD image showed that Dox was intercalated into montmorillonite, inter space of which increased to 31.3Å; the FT-IR spectra showed the vibration bands of PEI at 1 560 cm(-1) and 2 850 cm(-1), the vibration band of Dox at 1 350 cm(-1). Size analysis and SEM revealed that the size of nano-particle was 600 nm, and the zeta-potential was 30 mV. Drug release experiment explored that the nano-particle stably released drug in range of 6 X10(-4) ≊ 8 X10(-4) mg/ml within 72 h. MTT assay showed that the cell viability was over 80% in experiment condition in COS-7 and SKOV3 cell lines. 0.3 mg PEI-MMT nano-particle was able to protect gastric mucosa from alcohol. Multifunction system of PEI/Dox/MMT has been prepared successfully.
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
Sigma-Aldrich
Bentonite, CP
Sigma-Aldrich
Bentonite