Merck
  • Hexagonal comb cells of honeybees are not produced via a liquid equilibrium process.
Hexagonal comb cells of honeybees are not produced via a liquid equilibrium process.
Die Naturwissenschaften (2012-11-15)
Daniel Bauer, Kaspar Bienefeld
ABSTRACT
The nests of European honeybees (Apis mellifera) are organised into wax combs that contain many cells with a hexagonal structure. Many previous studies on comb-building behaviour have been made in order to understand how bees produce this geometrical structure; however, it still remains a mystery. Direct construction of hexagons by bees was suggested previously, while a recent hypothesis postulated the self-organised construction of hexagonal comb cell arrays; however, infrared and thermographic video observations of comb building in the present study failed to support the self-organisation hypothesis because bees were shown to be engaged in direct construction. Bees used their antennae, mandibles and legs in a regular sequence to manipulate the wax, while some bees supported their work by actively warming the wax. During the construction of hexagonal cells, the wax temperature was between 33.6 and 37.6 °C. This is well below 40 °C, i.e. the temperature at which wax is assumed to exist in the liquid equilibrium that is essential for self-organised building.
Sigma-Aldrich
Beeswax, refined
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Beeswax, bleached