Merck
  • Stereoselective synthesis of conjugated trienols from allylic alcohols and 1-iodo-1,3-dienes.
Stereoselective synthesis of conjugated trienols from allylic alcohols and 1-iodo-1,3-dienes.
Organic letters (2012-10-24)
Damien Brandt, Véronique Bellosta, Janine Cossy
ABSTRACT
The stereoselective synthesis of conjugated trienes has been achieved from allylic alcohols and 1-iodo-1,3-dienes using Pd(OAc)(2)/AgOAc.
Sigma-Aldrich
Allyl alcohol, ≥99%
Sigma-Aldrich
Allyl alcohol, ≥98.5%
Sigma-Aldrich
Allyl alcohol, ≥98.5%