Merck
  • Control of STN7 transcript abundance and transient STN7 dimerisation are involved in the regulation of STN7 activity.
Control of STN7 transcript abundance and transient STN7 dimerisation are involved in the regulation of STN7 activity.
Planta (2012-10-23)
Tobias Wunder, Qiuping Liu, Elena Aseeva, Vera Bonardi, Dario Leister, Mathias Pribil
ABSTRACT
Reversible phosphorylation of LHCII, the light-harvesting complex of photosystem II, controls its migration between the two photosystems (state transitions), and serves to adapt the photosynthetic machinery of plants and green algae to short-term changes in ambient light conditions. The thylakoid kinase STN7 is required for LHCII phosphorylation and state transitions in vascular plants. Regulation of STN7 levels occurs at the post-translational level, depends on the thylakoid redox state, and might involve reversible autophosphorylation. Here, we have analysed the effects of different light conditions and chemical inhibitors on the abundance of STN7 transcripts and their products. This analysis was performed in wild-type Arabidopsis thaliana plants, in several photosynthetic mutants, and in lines overexpressing STN7 (oeSTN7) or expressing mutant variants of STN7 carrying single or double cysteine-serine exchanges. It was found that accumulation of the STN7 protein is also controlled at the level of transcript abundance. Under certain conditions, exposure to high light or far-red light treatment, the relative decreases in LHCII phosphorylation can be attributed to decreases in STN7 abundance. Nevertheless, inhibitor experiments showed that redox control of LHCII kinase activity persists in oeSTN7 plants. STN7 dimers were found in oeSTN7 plants and in lines with single cysteine-serine exchanges, indicating that dimerisation involves disulphide bridges. We speculate that transient STN7 dimerisation is required for STN7 activity, and that the altered dimerisation behaviour of oeSTN7 plants might be responsible for the unusually high phosphorylation of LHCII in the dark found in this genotype.
Sigma-Aldrich
Diuron, ≥98%
Pricing and availability is not currently available.
Supelco
Diuron, PESTANAL®, analytical standard
Pricing and availability is not currently available.