Merck
  • Controlled release using a polymer stereocomplex capsule through the selective extraction and incorporation of one capsule shell component.
Controlled release using a polymer stereocomplex capsule through the selective extraction and incorporation of one capsule shell component.
Langmuir : the ACS journal of surfaces and colloids (2012-10-02)
Toshiyuki Kida, Masataka Mouri, Kenta Kondo, Mitsuru Akashi
ABSTRACT
Isotactic poly(methyl methacrylate) (it-PMMA)/syndiotactic poly(methacrylic acid) (st-PMAA) stereocomplex hollow capsules were fabricated by the deposition of stereocomplex films of it-PMMA and st-PMAA on silica particles by alternate layer-by-layer assembly and the subsequent removal of the silica particles with aqueous HF. The selective extraction of st-PMAA from the it-PMMA/st-PMAA stereocomplex capsule shells was successfully carried out by immersion in a pH 6-9 aqueous solution. The incorporation of st-PMAA into the resulting porous capsule shells was performed by immersion in an acetonitrile/water (1/1) solution of st-PMAA. The controlled release of an encapsulated dye from the it-PMMA/st-PMAA hollow capsules was achieved by combining the selective extraction of st-PMAA from the capsule shells and the incorporation of st-PMAA into the resulting porous shells.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mn ~5,400, average Mw ~9,500 by GPC, 30 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(methacrylic acid, sodium salt) solution, average Mw 4,000-6,000, 40 wt. % in H2O
Pricing and availability is not currently available.