Merck
  • Tibolone has anti-inflammatory effects in estrogen-deficient female rats on the natriuretic peptide system and TNF-alpha.
Tibolone has anti-inflammatory effects in estrogen-deficient female rats on the natriuretic peptide system and TNF-alpha.
Regulatory peptides (2012-09-15)
Ana Raquel Santos de Medeiros, Aline Zandonadi Lamas, Izabela Facco Caliman, Polyana L Meireles Dalpiaz, Luciana Barbosa Firmes, Gláucia Rodrigues de Abreu, Margareth Ribeiro Moysés, Elenice Moreira Lemos, Adelina Martha dos Reis, Nazaré Souza Bissoli
ABSTRACT
Cardiovascular and immune system abnormalities have been reported in females with estrogen deficiency. To control these disorders in post-menopausal women, hormone replacement therapy (HRT) has been used. Tibolone has been used as a HRT, but the effects of tibolone on the natriuretic peptide system have not been determined. We investigated the effects of tibolone on the natriuretic peptide system and pro-inflammatory cytokines in ovariectomized (OVX) rats. Female rats were divided into four groups: SHAM, OVX, OVX treated with 17β-estradiol (OVX+E: 14 days) and OVX treated with tibolone (OVX+T: 14 days) beginning 21 days after ovariectomy. On day 35, blood was collected to determine atrial natriuretic peptide (ANP), interleukin-6 (IL-6), and tumor necrosis factor-alpha (TNF-α) levels. In addition, tissues were collected for determining ANP, natriuretic peptide receptor type-A (NPR-A), and NPR type-C (NPR-C) gene expression levels by RT-PCR. The cytokine levels of both IL-6 and TNF-α were increased in OVX animals. In comparison, IL-6 and TNF-α levels were reduced in OVX+E animals. TNF-α levels were reduced similarly in OVX+T animals, but IL-6 levels remained elevated in this group. The concentrations of ANP in the left atrium tissue and plasma were decreased after ovariectomy, as were ANP mRNA levels in the left atrium and NPR-A mRNA levels in kidney. No variation in NPR-C gene expression in the kidney tissue was observed among the groups. Tibolone and 17β-estradiol effectively increased plasma ANP and ANP mRNA levels in the left atrium, but did not normalize renal NPR-A levels. Since HRT with tibolone normalizes plasma ANP and serum TNF-alpha levels our results suggest that treatment with tibolone has anti-inflammatory effects and could prevent cardiovascular disease in the long-term.