Merck
  • Effect of beta-alanine and carnosine supplementation on muscle contractility in mice.
Effect of beta-alanine and carnosine supplementation on muscle contractility in mice.
Medicine and science in sports and exercise (2012-08-17)
Inge Everaert, Sanne Stegen, Bert Vanheel, Youri Taes, Wim Derave
ABSTRACT
Enhanced carnosine levels have been shown to be ergogenic for high-intensity exercise performances, although the role of carnosine in the control of muscle function is poorly understood. Therefore, the aim of this study was to investigate the effect of long-term supplementation with increasing doses of carnosine and beta-alanine on muscle carnosine, anserine, and taurine levels and on in vitro contractility and fatigue in mice. Male Naval Medical Research Institute mice (n = 66) were control fed or supplemented with either carnosine (0.1%, 0.5%, or 1.8%) or beta-alanine (0.6 or 1.2%) in their drinking water for 8-12 wk. Soleus and extensor digitorum longus (EDL) were tested for in vitro contractile properties, and carnosine, anserine, and taurine content were measured in EDL and tibialis anterior by high-performance liquid chromatography. Only supplementation with 1.8% carnosine and 1.2% beta-alanine resulted in markedly higher carnosine (up to +160%) and anserine levels (up to +46%) compared with control mice. Beta-alanine supplementation (1.2%) resulted in increased fatigue resistance in the beginning of the fatigue protocol in soleus (+2%-4%) and a marked leftward shift of the force-frequency relation in EDL (10%-31% higher relative forces). Comparable with humans, beta-alanine availability seems to be the rate-limiting step for synthesis of muscle histidine-containing dipeptides in mice. Moreover, muscle histidine-containing dipeptides loading in mice moderately and muscle dependently affects excitation-contraction coupling and fatigue.
Sigma-Aldrich
L-Carnosine, ~99%, crystalline